


Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Boiling Pt:32…37,7 °C (1013 hPa)
Melting Pt:–60…–57,5 °C
Density:0,907 g/cm³ (20 °C)
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 25 %
- Evaporation residue:Max. 20 ppm
- Substances reducing KMnO4 (as O):Max. 8 ppm
- Total S (as SO4):Max. 1 ppm
- Cl (Chloride):Max. 0.5 ppm
- CO3 (as CO2):Max. 15 ppm
- PO4 (Phosphate):Max. 1 ppm
- Ca (Calcium):Max. 1 ppm
- Cd (Cadmium):Max. 0.1 ppm
- Cu (Copper):Max. 0.1 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 1 ppm
- Mn (Manganese):Max. 0.01 ppm
- Na (Sodium):Max. 2 ppm
- Pb (Lead):Max. 0.1 ppm
- Zn (Zinc):Max. 0.1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
sds_VWRC1133
SDS
Frequently Bought Together