Nitric acid 69%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Merck Index:13,06608
Boiling Pt:120,5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2031
ADR:8 (5.1),II
:M-Clarity™ quality level = MQ300
EINECS:231-714-2
:GHS03
:GHS06
:GHS05
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-41 °C
- Boiling Point:122 °C (1013 hPa)
- Vapor Pressure:9.4 hPa (20 °C)
- Density:1.41 g/cm3 (20 °C)
- pH:- 1 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.01799.2500_TDS_EN
Product specification
Frequently Bought Together