Order Entry
Ireland
ContactUsLinkComponent
Nitric acid 69% NF, J.T.Baker®
Oxidizing
Corrosive
Irritant
Nitric acid 69% NF, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Oxidizing
Corrosive
Irritant
Nitric acid 69% NF, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    69.0 - 70.0 %
  • NF - Assay (as HNO₃):
    69.0 - 70.0 %
  • NF - Clarity and Color:
    Passes Test
  • NF - Identification:
    Passes Test
  • NF - Chloride (Cl):
    ≤0.5 ppm
  • NF - Sulfate (SO₄):
    ≤1 ppm
  • NF - Iron (Fe):
    ≤0.2 ppm
  • NF - Residue on Ignition:
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together