Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
Supplier: Roth Carl
Certificates
About this item
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:3082
ADR:9,III
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together