Iodine 0.5 M I2 (1.0N) volumetric solution, Fluka™
35089-1L
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
- Factor:0.999 - 1.001
:
:253.81 g/mol
:>100 °C (1,013 hPa)
:1,260 g/cm³ (20 °C)
:Ambient
:231-442-4
:8,II
:GHS07
:GHS08
:7553-56-2
:I₂
:MFCD00011355