Order Entry
Ireland
ContactUsLinkComponent
Ammonium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution
Ammonium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # ROTHP718.1
Supplier:  Roth Carl
CAS Number:  
Ammonium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # ROTHP718.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :76.12 g/mol
Storage Temperature:Ambient
EINECS:217-175-6
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428