Hydrobromic acid 48% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Hydrogen bromide
CAS Number:10035-10-6
Formula:HBr
MDL Number:MFCD00011323
:80.91 g/mol
Merck Index:14,4778
Density:1,490
Storage Temperature:Ambient
EINECS:233-113-0
UN:1788
ADR:8,II
:GHS06
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together