



Sodium methoxide 25% in methanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:124-41-4
:
Formula:CH₃NaO
MDL Number:MFCD00012179
:54.02 g/mol
Density:0,97 g/cm³ (20 °C)
EINECS:204-699-5
UN:1289
ADR:3,III
:M-Clarity™ quality level = MQ200
:GHS02
:GHS06
:GHS05
:GHS08
Frequently Bought Together