Order Entry
Ireland
ContactUsLinkComponent
Linoleic acid ≥75% in aqueous solution, Technical Grade
Linoleic acid ≥75% in aqueous solution, Technical Grade
  A15265.18
 :  
Linoleic acid ≥75% in aqueous solution, Technical Grade
  A15265.18
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

 

 

 :60-33-3
 :
 :C₁₈H₃₂O₂
 :MFCD00064241
 :280.45 g/mol
 :14,5505
 :230 °C (16 mmHg)
 :0,902
 :>110 °C (230 °F)
 :Refrigerator
 :200-470-9