(±)-trans-1,2-Cyclohexanediol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1460-57-7
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00063611
Boiling Pt:231…233 °C
Melting Pt:101…106 °C
Density:1,147 g/cm³ (24 °C)
Flash Pt:134 °C (273 °F)
:116.16 g/mol
Storage Temperature:Ambient
EINECS:215-956-6
Related Information
Frequently Bought Together