Order Entry
Ireland
ContactUsLinkComponent
(±)-α-Methylbenzylamine for synthesis, Sigma-Aldrich®
Corrosive
Irritant
(±)-α-Methylbenzylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Corrosive
Irritant
(±)-α-Methylbenzylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    42 g/l (20 °C)
  • Melting Point:
    -65 °C
  • Molar Mass:
    121.18 g/mol
  • Boiling Point:
    184 - 186 °C (1013 hPa)
  • Vapor Pressure:
    0.7 hPa (20 °C)
  • Flash Point:
    70 °C
  • Refractive Index:
    1.5253 (20 °C, 589 nm)
  • Density:
    0.95 g/cm3 (20 °C)
  • pH:
    7 (H2O, 20 °C)
  • Ignition Point:
    355 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together