Order Entry
Ireland
ContactUsLinkComponent
N-Acetyl-DL-tryptophan 98+%, extra pure
N-Acetyl-DL-tryptophan 98+%, extra pure
Certificates
undefined
N-Acetyl-DL-tryptophan 98+%, extra pure
Specification Test Results
  • Appearance:
    White to beige Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 % (105°C)
  • Sulfated ash:
    ≤0.2 %
  • Titration excess NaOH:
    98.0 to 101.0 % (On dry substance)
  • Heavy metals (as Pb):
    ≤10 ppm