

(±)-2-Pentanol
Catalog # ROTH6824.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:88.15 g/mol
Merck Index:12,07258
Boiling Pt:118 °C (1013 hPa)
Melting Pt:–50 °C
Density:0,81 g/cm³ (20 °C)
Flash Pt:41 °C
Storage Temperature:Ambient
EINECS:227-907-6
UN:1105
ADR:3,III
:GHS02
:GHS07
CAS Number:6032-29-7
Formula:C₅H₁₂O
MDL Number:MFCD00004579