

(±)-2-Butanol, extra dry 99+%, AcroSeal®
:78-92-2
:
:H₃CCH₂CH(OH)CH₃
:MFCD00004569
:74.12 g/mol
:13,01540
:99 °C (760 mmHg)
:–115 °C
:0,81 g/cm³
:24 °C
:Ambient
:201-158-5
:1120
:3,III
:GHS07
:GHS02
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
- Residue after evaporation:≤0.002 %
Frequently Bought Together