(±)-3-Amino-1,2-propanediol for synthesis, Sigma-Aldrich®
Catalog # 8.18522.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 ml
- Seq:2315
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:91.11 g/mol
- Boiling Point:140 - 141 °C (12 hPa)
- Vapor Pressure:0.01 hPa (20 °C)
- Flash Point:186 °C
- Refractive Index:1.492 (20 °C, 589 nm)
- Density:1.19 g/cm3 (20 °C)
- pH:11 (100 g/l, H2O, 20 °C)
- Synonyms:1-Amino-2,3-propanediol
- Hill Formula:C3H9NO2
- Chemical Formula:NH2CH2CH(OH)CH2OH
Specification Test Results
About this item
:
:91.11 g/mol
Boiling Pt:264 °C (1013 hPa)
Density:1,1752 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:210-475-8
:GHS05
Storage Temperature:Ambient
CAS Number:616-30-8
Formula:C₃H₉NO₂
MDL Number:MFCD00008140