



3-Buten-2-one 94% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-94-4
:
Formula:C₄H₆O
MDL Number:MFCD00008777
:70.09 g/mol
Merck Index:12,06214
Boiling Pt:80 °C (760 mmHg)
Melting Pt:-7 °C
Density:0,864 g/cm³
Flash Pt:–7 °C
EINECS:201-160-6
UN:1251
ADR:6.1,I
:GHS06
:GHS05
:GHS09
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Water:≤5 %
- GC:≥93 %
- Stabilizer:≤0.7 % Acetonitrile
Specification Test Results
Frequently Bought Together