(±)-3-Chloro-1,2-propanediol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-24-2
:
Formula:C₃H₇ClO₂
MDL Number:MFCD00004712
:110.54 g/mol
Merck Index:14,2145
Boiling Pt:213 °C
Melting Pt:–40 °C
Density:1,322
Flash Pt:137 °C (278 °F)
EINECS:202-492-4
UN:2689
ADR:6.1,III
:GHS06
:GHS08
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together