Order Entry
Ireland
ContactUsLinkComponent
3-Chloro-1-iodopropane 98% stabilised
Irritant
3-Chloro-1-iodopropane 98% stabilised
Certificates
undefined
Irritant
3-Chloro-1-iodopropane 98% stabilised

Specification Test Results

  • Appearance:
    Clear colorless to yellow to orange to light brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5440 to 1.5490 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Stabilizer:
    Copper

Specification Test Results

Related Information

Frequently Bought Together