Order Entry
Ireland
ContactUsLinkComponent
(±)-3-Bromo-1,2-propanediol 97%
(±)-3-Bromo-1,2-propanediol 97%
Certificates
undefined
(±)-3-Bromo-1,2-propanediol 97%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5165 to 1.5195 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together