Order Entry
Ireland
ContactUsLinkComponent
 
Toxic
Health
Chloroform stabilised, EMPLURA®, Supelco®
Catalog # 8.22265.2500
Supplier:  Merck
CAS Number:  
undefined
Toxic
Health
Chloroform stabilised, EMPLURA®, Supelco®
Catalog # 8.22265.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :119.38 g/mol
Merck Index:13,02160
Boiling Pt:61,7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1,4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
REACH:05-2114614906-45
 :GHS06
 :GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826