Tannic acid, powder, EMPROVE® ESSENTIAL Ph. Eur., JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:1401-55-4
:85085-33-0
Formula:C₇₆H₅₂O₄₆
MDL Number:MFCD00066397
:1701.2 g/mol
Merck Index:12,09221
Melting Pt:218…219 °C
Density:>1 g/cm³ (20 °C)
Flash Pt:199 °C
EINECS:215-753-2
Related Information
Frequently Bought Together