Order Entry
Ireland
ContactUsLinkComponent
Tannic acid ≥96.0% (dry basis) FCC, Macron Fine Chemicals™
Tannic acid ≥96.0% (dry basis) FCC, Macron Fine Chemicals™
Catalog # 1674-21
CAS Number:  
undefined
Tannic acid ≥96.0% (dry basis) FCC, Macron Fine Chemicals™
Catalog # 1674-21
CAS Number:  

Specifications

  • Pack type:
    Box for dry chemical with liner
  • Pk:
    25 kg
  • Seq:
    4283
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Food GMP Manufactured Product:
    ≥96.0%
  • Meets F.C.C. Requirements:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Food Chemical:
    Passes Test
  • Assay () (dried basis) (FCC):
    ≥96.0 %
  • Appearance (fluffy, light tan to brown powder):
    Passes Test
  • Solubility:
    Passes Test
  • Gums or Dextrin:
    Passes Test
  • Resinous Substances:
    Passes Test
  • Identification A:
    Passes Test
  • Identification B:
    Passes Test
  • Loss on Drying at 105°C:
    ≤7.0 %
  • Residue on Ignition:
    ≤1.0 %
  • Lead (Pb):
    ≤2 mg/kg

Specification Test Results

About this item

Synonyms: Gallotannin, Tannin
 :
 :1701.2 g/mol
EINECS:215-753-2
CAS Number:1401-55-4
 :85085-33-0
Formula:C₇₆H₅₂O₄₆
MDL Number:MFCD00066397
Melting Pt:218…219 °C
Density:1 g/cm³ (20 °C)
Flash Pt:199
Merck Index:12,09221