Order Entry
Ireland
ContactUsLinkComponent
Tween® 40 (Polysorbate) for synthesis, Sigma-Aldrich®
Tween® 40 (Polysorbate) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Tween® 40 (Polysorbate) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Boiling Point:
    100 °C
  • Flash Point:
    149 °C
  • Density:
    1.09 g/cm3 (20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together