Calcium-D-gluconate monohydrate
Catalog # ROTH7502.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:448.39 g/mol
Melting Pt:178 °C
Storage Temperature:Ambient
EINECS:206-075-8
CAS Number:66905-23-5
:299-28-5
Formula:C₁₂H₂₂CaO₁₄·H₂O
MDL Number:MFCD00150787