Polyethylene glycol 4,000, scales, EMPROVE® ESSENTIAL Ph. Eur., SAFC®
8.17063.1000
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
:
:4000 g/mol
:∼250 °C (1013 hPa)
:53…58 °C
:1,13 g/cm³ (20 °C)
:250 °C
:Ambient
:500-038-2
:25322-68-3
:MFCD00081839
:H(OCH₂CH₂)nOH
:MFCD01779612