Order Entry
Ireland
ContactUsLinkComponent
Polyethylene glycol 2,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol 2,000 for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Polyethylene glycol 2,000 for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    600 g/l (20 °C)
  • Melting Point:
    49 - 52 °C
  • Molar Mass:
    1800.00 g/mol
  • Bulk Density:
    400 - 500 kg/m3
  • Flash Point:
    250 °C
  • Density:
    1.21 g/cm3 (20 °C)
  • pH:
    4 - 7 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    420 °C
  • Viscosity kinematic:
    46 - 55 mm2/s (20 °C) (50% solution)

Specification Test Results

Related Information

Frequently Bought Together