0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Proteins and Peptides
Colistin sulphate
Colistin sulphate
A2922.0001
:
PanReac AppliChem
:
Colistin sulphate
A2922.0001
:
PanReac AppliChem
:
Pack type:
Glass bottle for solids
Pk:
1 g
CAS No.:
1264-72-8
Storage conditions:
Refrigerator
Protein/peptide name:
Colistin sulphate