
Canada balsam, natural filtered
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:8007-47-4
:
MDL Number:MFCD00132800
Merck Index:14,946
Density:0,99
Flash Pt:43 °C (109 °F)
Storage Temperature:Ambient
EINECS:232-362-2
UN:1993
ADR:3,III
:GHS02
Related Information
Frequently Bought Together