Starch (from wheat), Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:9005-25-8
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
Density:1,5 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:162.14 g/mol
Storage Temperature:Ambient
EINECS:232-679-6
Related Information
Frequently Bought Together