Starch, soluble, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Starch, Vitex® iodine indicator, Amylodextrin
CAS Number:9005-84-9
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
Molecular Weight:162.14 g/mol
Merck Index:13,08878
Storage Temperature:Ambient
EINECS:232-686-4
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White fine powder
- pH (2 %):5,5 - 7,5
- Sensitivity:Passes test
- Ignition residue:Max. 0.4 %
- Loss on drying (105°C; 2 h):Max. 10 %
- Reducing substances (as C12H22O11):Max. 0.7 %
Specification Test Results
Related Information
Frequently Bought Together