Agarose LM, low melting for recovery of samples after separation
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9012-36-6
:
MDL Number:MFCD00081294
Melting Pt:60…90 °C
Storage Temperature:Ambient
EINECS:232-731-8
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together