Cellulose, Avicel
Supplier: MACHEREY-NAGEL
Certificates
About this item
CAS Number:9004-34-6
:
Formula:(C₁₂H₂₂O₁₁)n
MDL Number:MFCD00081512
:342.3 g/mol
Merck Index:13,01977
Melting Pt:260…270 °C
Density:1,27…1.6 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-674-9
Frequently Bought Together