
Gentamicin sulphate
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:1405-41-0
:
MDL Number:MFCD00270181
Merck Index:13,04403
Melting Pt:218…237 °C (lit.)
EINECS:215-778-9
:GHS08
Storage Temperature:Ambient
Frequently Bought Together