D-(+)-Sucrose ≥99.9%, Ultrapure DNase-, RNase-free
:57-50-1
:
:C₁₂H₂₂O₁₁
:MFCD00006626
:13,08966
:169…170 °C
:1,29 g/cm³ (20 °C)
:342.3 g/mol
:Ambient
:200-334-9
- Chloride:≤0.005%
- DNase:None detected
- Glucose:≤0.1%
- Heavy Metals (as Pb):≤0.0005%
- Insolubles:≤0.005%
- Invert Sugar:≤0.1%
- Iron:≤0.0005%
- Loss on Drying:≤0.03%
- Protease:none detected
- Purity:≤99.9%
- RNase:none detected
- Solubility (10%, Water):Pass
- Specific Rotation:66.3 – 66.8°
- Sulphate:≤0.005%
Frequently Bought Together