D-(+)-Sucrose ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:57-50-1
:
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006626
Merck Index:14,8881
Melting Pt:186…190 °C
Density:1,580
:342.3 g/mol
Storage Temperature:Ambient
EINECS:200-334-9
Related Information
Frequently Bought Together