Toluene-[D₈]
Catalog # ROTHCP95.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:100.08 g/mol
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–85 °C
Density:0,94 g/cm³ (20 °C)
Flash Pt:4 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:218-009-5
UN:1294
ADR:3,II
:GHS02
:GHS08
:GHS07
CAS Number:2037-26-5
Formula:C₆D₅CD₃
MDL Number:MFCD00044638