Acetone-[D₆] 99.5 atom % D + 1% TMS for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:666-52-4
:
Formula:D₃CC(O)CD₃
MDL Number:MFCD00044635
:64.03 g/mol
Boiling Pt:55,5 °C (1013 hPa)
Melting Pt:–93 °C
Density:0,87 g/cm³
Flash Pt:–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:211-563-9
UN:1090
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 % (Coulometric)
- D-Enrichment:≥99.4 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together