Order Entry
Ireland
ContactUsLinkComponent
 
Acetone-[D₆] 99.9 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
 
undefined
Acetone-[D₆] 99.9 atom % D + 0.03% (v/v) TMS for NMR spectroscopy

 

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.03 % (Coulometric)
  • D-Enrichment:
    ≥99.85 %

 

 

Frequently Bought Together