Petroleum spirit, 40…60 °C, ECD tested for residue analysis lindane/DDT <5ng/l, res <0.0003%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Gasoline, Petroleum ether, Petrol, Benzine
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:40…60 °C (1013 hPa)
Density:0,65 g/cm³
Flash Pt:–45 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
:GHS08
:GHS09
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.005 % (Coulometric)
- Boiling point:40 to 60 °C
- Color scale:≤10 APHA
- GC/ECD (as lindane):≤5 ng/l
- Residue after evaporation:≤0.0003 %
- Acidity (CH3COOH):≤0.003 %
Specification Test Results
Frequently Bought Together