Molecular sieve 3A (0.3 nm, 3 Å), pellets, Ø 1 - 2 mm (0.04 - 0.08 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:308080-99-1
:1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00131613
Storage Temperature:Ambient
Frequently Bought Together