Order Entry
Ireland
ContactUsLinkComponent
 
Molecular sieve 3A (0.3 nm, 3 Å) 8-12 mesh
 
undefined
Molecular sieve 3A (0.3 nm, 3 Å) 8-12 mesh

 

  • Appearance:
    Beige to light brown Beads
  • Particle size:
    8 to 12 mesh
  • Bulk density:
    ≥600 g/l
  • Residual water:
    ≤1.5 %
  • Water adsorption:
    ≥20 % (60% rH)

 

Frequently Bought Together