Order Entry
Ireland
ContactUsLinkComponent
 
Palladium ≥99%, powder, EMSURE® for analysis, Supelco®
 :  Merck
 
Palladium ≥99%, powder, EMSURE® for analysis, Supelco®
 :  Merck

 

  • Environmentally Preferable:

 

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    1554 °C
  • Molar Mass:
    106.4 g/mol
  • Boiling Point:
    2200 - 2900 °C
  • Density:
    12.02 g/cm3 (20 °C)
  • pH:
    (H2O) not applicable

 

Frequently Bought Together