Order Entry
Ireland
ContactUsLinkComponent
Nickel, powder, Sigma-Aldrich®
Health
Irritant
Nickel, powder, Sigma-Aldrich®
Supplier:  Merck
Certificates
Health
Irritant
Nickel, powder, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    1453 °C
  • Molar Mass:
    58.69 g/mol
  • Bulk Density:
    1600 - 2600 kg/m3
  • Boiling Point:
    2832 °C
  • Density:
    8.90 g/cm3 (25 °C)

Specification Test Results

Related Information

Frequently Bought Together