Molybdenum, powder, Sigma-Aldrich®
Catalog # 1.12254.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Hill Formula:Mo
- Molar Mass:95.94 g/mol
- Density:10.2 g/cm³ (20 °C)
- Melting Point:2620 °C
- Bulk density:1400 kg/m³
- Solubility:(20 °C) insoluble
- Storage class:10 - 13 Other liquids and solids
- WGK:NWG not water endangering
Specification Test Results
About this item
:
Boiling Pt:5560 °C (1013 hPa)
Melting Pt:2622 °C
Density:10,2 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3089
ADR:4.1,III
:M-Clarity™ quality level = MQ200
:95.94 g/mol
EINECS:231-107-2
CAS Number:7439-98-7
Formula:Mo
MDL Number:MFCD00003465