Jump to:
Product Details & Documents
- CAS Number:7440-62-2
- Formula:V
- MDL Number:MFCD00011453
- Molecular Weight:50.94 g/mol
- Density:6,11 g/cm³ (187 °C)
- Storage Temperature:Ambient
- EINECS:231-171-1
- UN:3089
- ADR:4.1,II
- :M-Clarity™ quality level = MQ200
Documents
msds_112392
SDS
ProductPage_112392
Product specification
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:1710 °C
- Molar Mass:50.94 g/mol
- Bulk Density:4000 kg/m3
- Boiling Point:3000 °C
- Density:6.11 g/cm3 (18.7 °C)
Specifications
- Pack type:Glass bottle
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



