Gold ≥99.9% (metals basis), thin foil, Thickness: 0.006 mm (0.00024 in)
Catalog # 44138.FF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1643
- Pk Info:25×25 mm
Specifications
About this item
CAS Number:7440-57-5
:
Formula:Au
MDL Number:MFCD00003436
:196.97 g/mol
Merck Index:12,04526
Boiling Pt:2800 °C (1013 hPa)
Melting Pt:1063 °C
Density:19,3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-165-9