Order Entry
Ireland
ContactUsLinkComponent
Gold ≥99.9% (metals basis), thin foil, Thickness: 0.003 mm (0.00012 in)
Gold ≥99.9% (metals basis), thin foil, Thickness: 0.003 mm (0.00012 in)
Catalog # 44137.FI
CAS Number:  
Gold ≥99.9% (metals basis), thin foil, Thickness: 0.003 mm (0.00012 in)
Catalog # 44137.FI
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1670
  • Pk Info:
    50×50 mm

Specifications

About this item

CAS Number:7440-57-5
 :
Formula:Au
MDL Number:MFCD00003436
 :196.97 g/mol
Merck Index:12,04526
Boiling Pt:2800 °C (1013 hPa)
Melting Pt:1063 °C
Density:19,3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-165-9