Gold ≥99.999% (metals basis), wire, Premion®, Ø 0.762 mm (0.030 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-57-5
:
Formula:Au
MDL Number:MFCD00003436
Boiling Pt:2800
Melting Pt:1063
Density:19.3 g/cm³ (20 °C)
Molecular Weight:196.97 g/mol
Merck Index:12,04526
Storage Temperature:Ambient
EINECS:231-165-9
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together