

Sulphur ≥99.5%, powder -325 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7704-34-9
:
Formula:S
MDL Number:MFCD00085316
:32.07 g/mol
Merck Index:13,09059
Boiling Pt:445 °C (1013 hPa)
Melting Pt:113…119 °C
Density:2,36 g/cm³ (20 °C)
Flash Pt:168 °C
Storage Temperature:Ambient
EINECS:231-722-6
UN:1350
ADR:4.1,III
:GHS02
:GHS07
Related Information
Frequently Bought Together