Bismuth ≥99.997% (metals basis), granules 1-2 mm (0.04-0.08 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-69-9
:
Formula:Bi
MDL Number:MFCD00134033
:208.98 g/mol
Merck Index:12,01297
Boiling Pt:1490 °C (1013 hPa)
Melting Pt:271…272 °C
Density:9,78 g/cm³ (20 °C)
:Packaged under argon.
Storage Temperature:Ambient
EINECS:231-177-4
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together